missing translation for 'onlineSavingsMsg'
Learn More

DL-Ornithine monohydrochloride, 99%

Product Code. 11408960
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11408960 50 g 50g
11423697 5 g 5g
11433697 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11408960 Supplier Thermo Scientific Alfa Aesar Supplier No. A18173.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1069-31-4 | C5H13ClN2O2 | 168.62 g/mol

Erythro-3-hydroxy-DL-ornithine monohydrochloride is used as a reagent to synthesize DL-Capreomycidine, a guanidine (HCl)-containing amino acid that is a component of antituberculous peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Erythro-3-hydroxy-DL-ornithine monohydrochloride is used as a reagent to synthesize DL-Capreomycidine, a guanidine (HCl)-containing amino acid that is a component of antituberculous peptides.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1069-31-4
Molecular Formula C5H13ClN2O2
Molecular Weight (g/mol) 168.62
MDL Number MFCD00065398
InChI Key GGTYBZJRPHEQDG-UHFFFAOYNA-N
Synonym dl-ornithine hydrochloride, 2,5-diaminopentanoic acid hydrochloride, dl-ornithine monohydrochloride, dl-ornithinehcl, h-dl-orn-oh.hcl, dl-ornithine hcl, dl-ornithine, hydrochloride, ornithine monohydrochloride, ornithine hydrochloride, ornithine, hydrochloride 1:1
PubChem CID 71598
IUPAC Name hydrogen 2,5-diaminopentanoic acid chloride
SMILES [H+].[Cl-].NCCCC(N)C(O)=O

Specifications

Melting Point ∼225°C (decomposition)
Beilstein 4153338
Solubility Information Soluble in water.
Quantity 50 g
Formula Weight 168.62
Percent Purity 99%
Chemical Name or Material DL-Ornithine monohydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.