missing translation for 'onlineSavingsMsg'
Learn More

DL-Pantolactone, 97%

Product Code. 10756495 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10756495 25 g Glass bottle 25g
10507644 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10756495 Supplier Thermo Scientific Acros Supplier No. 427590250
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 79-50-5 | C6H10O3 | 130.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-50-5
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
MDL Number MFCD00064333
InChI Key SERHXTVXHNVDKA-UHFFFAOYSA-N
Synonym pantolactone, dl-pantolactone, 3-hydroxy-4,4-dimethyldihydrofuran-2 3h-one, pantoyl lactone, pantoic lactone, dl-pantoyl lactone, d---pantolactone, 2 3h-furanone, dihydro-3-hydroxy-4,4-dimethyl, dihydro-3-hydroxy-4,4-dimethyl-2 3h-furanone, pantolyl lactone
PubChem CID 989
IUPAC Name 3-hydroxy-4,4-dimethyloxolan-2-one
SMILES CC1(COC(=O)C1O)C

Specifications

Melting Point 74.0°C to 78.0°C
Boiling Point 246.0°C
Flash Point 122°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: soluble.
Formula Weight 130.14
Percent Purity 97%
Chemical Name or Material DL-Pantolactone
compliance-icons
Product Identifier
  • DL-Pantolactone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.