missing translation for 'onlineSavingsMsg'
Learn More

DL-Phenylalanine, ≥99% (Dry Basis), Thermo Scientific™

Product Code. 15875328 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15875328 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15875328 Supplier Thermo Scientific Supplier No. J20100.A1
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 150-30-1
Physical Form Crystalline
Quantity 1 kg
Molecular Formula C9H11NO2
MDL Number MFCD00064225
Packaging Plastic Bottle
Synonym dl-phenylalanine
InChI Key COLNVLDHVKWLRT-UHFFFAOYNA-N
SMILES NC(CC1=CC=CC=C1)C(O)=O
IUPAC Name 2-amino-3-phenylpropanoic acid
Molecular Weight (g/mol) 165.192
Chemical Name or Material DL-Phenylalanine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.