missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Propranolol hydrochloride, 99%

Product Code. 10462401 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10462401

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 318-98-9 | C16H21NO2·HCl | 295.8 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 318-98-9
Molecular Formula C16H21NO2·HCl
Molecular Weight (g/mol) 295.8
MDL Number MFCD00012558
InChI Key ZMRUPTIKESYGQW-UHFFFAOYSA-N
Synonym propranolol hydrochloride, propranolol hcl, inderal, avlocardyl, herzbase, inderalici, naprilin, pronovan, dociton, ikopal
PubChem CID 62882
ChEBI CHEBI:8500
IUPAC Name 1-naphthalen-1-yloxy-3-(propan-2-ylamino)propan-2-ol;hydrochloride
SMILES CC(C)NCC(COC1=CC=CC2=CC=CC=C21)O.Cl

Specifications

Melting Point 163.0°C to 166.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C10H7OCH2CH(OH)CH2NHCH(CH3)2·HCl
Quantity 5 g
Merck Index 15, 7953
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, practically insoluble in ether, benzene and ethyl, acetate
Formula Weight 295.8
Percent Purity 99%
Physical Form Powder
Chemical Name or Material DL-Propranolol hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • DL-Propranolol hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.