missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Serine, 99%

Product Code. 10592911 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10592911 100 g Glass Bottle 100g
10336730 25 g Glass Bottle 25g
10083853 500 g Glass Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10592911 Supplier Thermo Scientific Chemicals Supplier No. 132651000
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 302-84-1 | C3H7NO3 | 105.093 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 302-84-1
Molecular Formula C3H7NO3
Molecular Weight (g/mol) 105.093
MDL Number MFCD00064223
InChI Key MTCFGRXMJLQNBG-UHFFFAOYSA-N
Synonym dl-serine, serin, serine dl-form, h-dl-ser-oh, 3-hydroxyalanine, serine, l, serine, dl, h-d-ser-oh, dl-2-amino-3-hydroxypropionic acid
PubChem CID 617
ChEBI CHEBI:17822
IUPAC Name 2-amino-3-hydroxypropanoic acid
SMILES C(C(C(=O)O)N)O

Specifications

Ammonium 200ppm max.
Color White
Chlorine (Cl) 200ppm max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 10ppm max.
Assay Percent Range 99%
Loss on Drying 0.3% max. (105°C, 3 hrs)
Other Amino Acids 0.5% max. (TLC)
Packaging Glass Bottle
Linear Formula HOCH2CH(NH2)CO2H
Beilstein 04, 511
Sulfate 500ppm max.
Merck Index 15, 8599
Transmittance 95% min. (c=10, 2N HCl, 430nm) 1cm cell
Sulfated Ash 0.1% max.
Density 1.53g/mL
Quantity 100 g
Formula Weight 105.09
Percent Purity 98.5 to 101.5%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material DL-Serine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.