missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Serine, 99%

Product Code. 11492188
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11492188 500 g 500g
11482188 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11492188 Supplier Thermo Scientific Chemicals Supplier No. A15184.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 302-84-1 | C3H7NO3 | 105.093 g/mol

DL-Serine is used in the synthesis of novel tryptoline derivatives as IDO (indoleamine 2,3-Deoxygenase) inhibitors, for potential use in Alzheimer’s treatment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Serine is used in the synthesis of novel tryptoline derivatives as IDO (indoleamine 2,3-Deoxygenase) inhibitors, for potential use in Alzheimer’s treatment.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 302-84-1
Molecular Formula C3H7NO3
Molecular Weight (g/mol) 105.093
MDL Number MFCD00064223
InChI Key MTCFGRXMJLQNBG-UHFFFAOYSA-N
Synonym dl-serine, serin, serine dl-form, h-dl-ser-oh, 3-hydroxyalanine, serine, l, serine, dl, h-d-ser-oh, dl-2-amino-3-hydroxypropionic acid
PubChem CID 617
ChEBI CHEBI:17822
IUPAC Name 2-amino-3-hydroxypropanoic acid
SMILES C(C(C(=O)O)N)O

Specifications

Melting Point ∼240°C (decomposition)
Beilstein 1721402
Merck Index 14,8460
Solubility Information Soluble in water.
Quantity 500 g
Formula Weight 105.09
Percent Purity 99%
Chemical Name or Material DL-Serine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.