Learn More
Abcam DL-Serine Assay Kit
Product Code. 32120221 Shop All Abcam Products
Change view
Click to view available options
100 Tests
Product Code.
32120221 100 Tests
1 options
Product Code. 32120221 Supplier Abcam Supplier No. ab241027100Test
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Measure D Serine in 1h 15m in Biofluids, Cell/Tissue Extracts with DL-Serine Assay Kit AB241027. Sensitivity <0.1 nmol/well. Cited in 8 publications. Use with Microplate reader.
Measure D Serine in 1h 15m in Biofluids, Cell/Tissue Extracts with DL-Serine Assay Kit AB241027. Sensitivity <0.1 nmol/well. Cited in 8 publications. Use with Microplate reader.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 100 Tests