missing translation for 'onlineSavingsMsg'
Learn More

DL-Tartaric acid, 99.5%

Product Code. 10676924 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10676924 100 g Plastic bottle 100g
10487330 500 g Plastic bottle 500g
11915061 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10676924 Supplier Thermo Scientific Acros Supplier No. 137871000
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 133-37-9 | C4H6O6 | 150.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 133-37-9
Molecular Formula C4H6O6
Molecular Weight (g/mol) 150.09
MDL Number MFCD00071626
InChI Key FEWJPZIEWOKRBE-UHFFFAOYNA-N
Synonym dl-tartaric acid, 2,3-dihydroxysuccinic acid, tartaric acid, racemic acid, uvic acid, traubensaure, paratartaric acid, paratartaric aicd, dl-tartrate, racemic tartaric acid
PubChem CID 875
ChEBI CHEBI:15674
IUPAC Name 2,3-dihydroxybutanedioic acid
SMILES OC(C(O)C(O)=O)C(O)=O

Spezifikation

Melting Point 200.0°C to 206.0°C
Color White
Quantity 100 g
Flash Point 210°C
Infrared Spectrum Authentic
Loss on Drying 0.5% max.
Packaging Plastic bottle
Physical Form Crystalline Powder
Assay Percent Range 99.50%
Linear Formula HO2CCH(OH)CH(OH)CO2H
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and ether, insoluble in chloroform
Formula Weight 150.09
Percent Purity 99.5%
Chemical Name or Material DL-Tartaric acid
Mehr anzeigen Weniger anzeigen
compliance-icons
Produktidentifikator
  • DL-Tartaric acid
Signalwort
  • Danger
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 1
Gefahrenhinweis
  • H318-Causes serious eye damage.
Sicherheitshinweis
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.