missing translation for 'onlineSavingsMsg'
Learn More

Dl-Tartaric Acid, TraceSELECT™, ≥99.9995% (Metals basis), Solstice

Product Code. 15631200 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Poly bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15631200 25 g Poly bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15631200 Supplier Honeywell-Fluka Supplier No. 1281925G
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 133-37-9
Melting Point 210°C to 212°C
Molecular Formula C4H6O6
Linear Formula HOOC(CHOH)2COOH
MDL Number MFCD00071626
UN Number NONH for all modes of transport
Beilstein 1725148
Synonym dl-tartaric acid, 2,3-dihydroxysuccinic acid, tartaric acid, racemic acid, uvic acid, traubensaure, paratartaric acid, paratartaric aicd, dl-tartrate, racemic tartaric acid
InChI Key FEWJPZIEWOKRBE-UHFFFAOYNA-N
SMILES OC(C(O)C(O)=O)C(O)=O
IUPAC Name 2,3-dihydroxybutanedioic acid
Molecular Weight (g/mol) 150.09
PubChem CID 875
ChEBI CHEBI:15674
Formula Weight 150.09g/mol
Percent Purity ≥99.9995% (metals basis)
Grade TraceSELECT
Quantity 25 g
Flash Point 210°C (410°F)
Packaging Poly bottle
Physical Form Solid
Chemical Name or Material Dl-Tartaric Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.