missing translation for 'onlineSavingsMsg'
Learn More

DL-Tetrahydropalmatine, TRC

Product Code. 30412511
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30412511 100 mg
30409076 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30412511 Supplier Toronto Research Chemicals Supplier No. TRCT294200100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2934-97-6
Molecular Formula C21 H25 N O4
Molecular Weight (g/mol) 355.43
Synonym 6H-Dibenzo[a,g]quinolizine, 5,8,13,13a-tetrahydro-2,3,9,10-tetramethoxy-, 6H-Dibenzo[a,g]quinolizine, 5,8,13,13a-tetrahydro-2,3,9,10-tetramethoxy-, (±)-, Berbine, 2,3,9,10-tetramethoxy-, (±)- (8CI), DL-Berbine, 2,3,9,10-tetramethoxy- (6CI), (R,S)-Tetrahydropalmatine, (±)-Corydalis B, (±)-Tetrahydropalmatine, 5,8,13,13a-Tetrahydro-2,3,9,10-tetramethoxy-6H-dibenzo[a,g]quinolizine, DL-Tetrahydropalmatine, THP, Tetrahydropalmatine, dl-Tetrahydropalmatine
IUPAC Name 2,3,9,10-tetramethoxy-6,8,13,13a-tetrahydro-5H-isoquinolino[3,2-a]isoquinoline
SMILES COc1cc2CCN3Cc4c(CC3c2cc1OC)ccc(OC)c4OC

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C21H25NO4/c1-23-18-6-5-13-9-17-15-11-20(25-3)19(24-2)10-14(15)7-8-22(17)12-16(13)21(18)26-4/h5-6,10-11,17H,7-9,12H2,1-4H3
Formula Weight 355.1784
Chemical Name or Material DL-Tetrahydropalmatine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.