missing translation for 'onlineSavingsMsg'
Learn More

DL-Thioctic acid, 98%, Thermo Scientific™

DL-Thioctic acid, CAS # 1077-28-7, is an antioxidant and coenzyme important for the activity of enzymes.

68.70 € - 222.00 €

Chemical Identifiers

CAS 1077-28-7
Molecular Formula C8H14O2S2
Molecular Weight (g/mol) 206.32
MDL Number MFCD00005474
InChI Key AGBQKNBQESQNJD-UHFFFAOYNA-N
Synonym dl-thioctic acid, thioctic acid, alpha-lipoic acid, 5-1,2-dithiolan-3-yl pentanoic acid, dl-alpha-lipoic acid, 1,2-dithiolane-3-pentanoic acid, 6,8-thioctic acid, thioctacid, 6-thioctic acid, alpha lipoic acid
PubChem CID 864
ChEBI CHEBI:16494
IUPAC Name 5-(1,2-dithiolan-3-yl)pentanoic acid
SMILES OC(=O)CCCCC1CCSS1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11321687
SDS View Documents
Thermo Scientific Alfa Aesar
L04711.06
5g
68.70 €
5g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
11409820
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
L04711.14
25g
222.00 €
25g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

General Description

• DL-Thioctic acid derives from octanoic acid, and it has been described as an antioxidant coenzyme important for the activity of enzyme complexes including pyruvate dehydrogenase and glycine decarboxylase

Application

• DL-Thioctic acid has been used in in vivo studies to increase insulin sensitivity and stimulate glucose uptake into adipose and muscle cells in vitro

• DL-Thioctic acid has an essential role in aerobic metabolism

• It has been used as free radical scavenger compound

• It can also assist in repairing oxidative damage via the increase of endogenous antioxidants, including vitamins C, vitamin E, and glutathione

• It can induce apoptosis through caspase-independent and caspase-dependent pathways, a process mediated by intracellular Ca2+

• DL-Thioctic acid has the ability to impact various cellular biological pathways since it can increase glutathione synthesis and modulate transcription factor activity, mainly nuclear factor kappa B (NF-κB)

• It can decrease the phagocytosis of myelin by macrophages

Specifications

Chemical Identifiers

1077-28-7
206.32
AGBQKNBQESQNJD-UHFFFAOYNA-N
864
5-(1,2-dithiolan-3-yl)pentanoic acid
C8H14O2S2
MFCD00005474
dl-thioctic acid, thioctic acid, alpha-lipoic acid, 5-1,2-dithiolan-3-yl pentanoic acid, dl-alpha-lipoic acid, 1,2-dithiolane-3-pentanoic acid, 6,8-thioctic acid, thioctacid, 6-thioctic acid, alpha lipoic acid
CHEBI:16494
OC(=O)CCCCC1CCSS1

Specifications

1077-28-7
C8H14O2S2
81853
dl-thioctic acid, thioctic acid, alpha-lipoic acid, 5-1,2-dithiolan-3-yl pentanoic acid, dl-alpha-lipoic acid, 1,2-dithiolane-3-pentanoic acid, 6,8-thioctic acid, thioctacid, 6-thioctic acid, alpha lipoic acid
AGBQKNBQESQNJD-UHFFFAOYNA-N
5-(1,2-dithiolan-3-yl)pentanoic acid
864
206.33
DL-Thioctic acid
160°C to 165°C
MFCD00005474
14,9326
Soluble in methanol,ethanol,diethyl ether and chloroform. Insoluble in water.
OC(=O)CCCCC1CCSS1
206.32
CHEBI:16494
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'einecsNumber' : 214-071-2

missing translation for 'rtecsNumber' : JP1192000

missing translation for 'tsca' : No

Recommended Storage : Keep cold

Product Identifier
  • DL-Thioctic acid

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contain: DL-Thioctic acid

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.