missing translation for 'onlineSavingsMsg'
Learn More

DL-Thiorphan, Thermo Scientific™

 

Brand:  Thermo Scientific Alfa Aesar J62557.MB

Quantity 25mg

Product Code. 15404919

  • ONLINE EXCLUSIVE PRICE
    448.00 €
    439.00 € / 25mg
    Save 9.00 €
    2% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Thiorphan has the ability to block the degradation of the luteinizing hormone releasing hormone (LHRH). At higher concentrations, (14 μM) this compound has been shown to inhibit the activity of angiotensin-converting enzyme (ACE). N
TRUSTED_SUSTAINABILITY
Specifications

Specifications

76721-89-6
MFCD00058435
4872101
Soluble in ethanol at 50mg/ml
OC(=O)CNC(=O)C(CS)CC1=CC=CC=C1
253.32
CHEBI:9568
DL-Thiorphan
C12H15NO3S
25mg
thiorphan, dl-thiorphan, dl-3-mercapto-2-benzylpropanoylglycine, n-2-benzyl-3-sulfanylpropanoyl glycine, chembl10247, glycine, n-2-mercaptomethyl-1-oxo-3-phenylpropyl-, +-, 2-2-benzyl-3-sulfanylpropanoyl amino acetic acid, 2-2-benzyl-3-mercaptopropanamido acetic acid, n-rs-2-benzyl-3-mercaptopropanoyl-glycine, tiorphan
LJJKNPQAGWVLDQ-UHFFFAOYNA-N
2-(2-benzyl-3-sulfanylpropanamido)acetic acid
3132
253.32
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.