missing translation for 'onlineSavingsMsg'
Learn More

DL-α-Tocopherol acetate, MedChemExpress

Product Code. 16448716
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16448716 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16448716 Supplier Medchem Express Supplier No. HYB1278A
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress DL-α-Tocopherol acetate is a vitamin E derivative which is often included in the formulations of enteral nutrition.

DL-α-Tocopherol acetate is a vitamin E derivative which is often included in the formulations of enteral nutrition.

Specifications

Chemical Name or Material DL-α-Tocopherol acetate
CAS 52225-20-4
Color Colorless
Purity Grade Notes Research
Molecular Formula C31H52O3
Quantity 100 mg
Synonym Vitamin E acetate
Solubility Information DMSO : ≥ 100 mg/mL (211.53 mM)
SMILES CC(CCC[C@H](CCC[C@H](CCC[C@@]1(CCC2=C(C(OC(C)=O)=C(C(C)=C2O1)C)C)C)C)C)C.CC(CCC[C@H](C)CCC[C@H](C)CCC[C@@]3(C)CCC4=C(C(OC(C)=O)=C(C(C)=C4O3)C)C)C
Molecular Weight (g/mol) 472.74
Formula Weight 472.74
Percent Purity 98.0%
Grade Research
Physical Form Oil
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.