missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Tryptophan, 99%

Product Code. 11322296
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11322296 5 g 5g
11332296 25 g 25g
11342296 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11322296 Supplier Thermo Scientific Chemicals Supplier No. L05936.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

DL-Tryptophan, CAS # 54-12-6, is an α-amino acid that is commonly used in the synthesis of proteins. It is an essential amino acid precursor for the formation of serotonin, melatonin, and vitamin B3. | CAS: 54-12-6 | C11H12N2O2 | 204.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

General Description

• DL-Tryptophan is an α-amino acid, and an essential amino acid precursor of serotonin, melatonin, and vitamin B3

Application

• DL-Tryptophan shows antioxidant activity• It has a role as a Daphnia magna metabolite

• It has been explored in in vitro studies to evaluate the antimicrobial and immunoregulatory functions on indoleamine 2,3-dioxygenase

• It has been used as a medium supplement in Psilocybe atrobrunnea and Psilocybe serbica for the synthesis of compounds

• It can be used as a medium supplement for culturing yeast

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54-12-6
Molecular Formula C11H12N2O2
Molecular Weight (g/mol) 204.23
MDL Number MFCD00064339
InChI Key QIVBCDIJIAJPQS-UHFFFAOYSA-N
Synonym dl-tryptophan, 2-amino-3-1h-indol-3-yl propanoic acid, racemic tryptophan, dl-trytophane, dl-trytophan, +--tryptophan, h-dl-trp-oh, dl-3beta-indolylalanine, dl-tryptophane, tryptophan .
PubChem CID 1148
ChEBI CHEBI:27897
IUPAC Name 2-amino-3-(1H-indol-3-yl)propanoic acid
SMILES NC(CC1=CNC2=CC=CC=C12)C(O)=O

Specifications

Melting Point ∽280°C (decomposition)
Beilstein 86196
Solubility Information Soluble I water. 10g/L (20°C),Hcl and alkalies. Sparingly soluble in alcohol.
Quantity 5 g
Formula Weight 204.23
Percent Purity 99%
Chemical Name or Material DL-Tryptophan

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.