missing translation for 'onlineSavingsMsg'
Learn More

DL-Valine methyl ester hydrochloride, 99%

Product Code. 11322556
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11322556 25 g 25g
11312556 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11322556 Supplier Thermo Scientific Alfa Aesar Supplier No. L06526.14
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 5619-05-6 | C9H12O2 | 152.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5619-05-6
MDL Number MFCD00066118

Specifications

Melting Point 112°C to 114°C
Molecular Formula C6H14ClNO2
Beilstein 3912090
Synonym dl-valine methyl ester hydrochloride, h-dl-val-ome.hcl, methyl 2-amino-3-methylbutanoate hydrochloride, h-d-val-ome.hcl, methyl dl-valinate hydrochloride, d,l-valine methyl ester hydrochloride, h-d-val-ome*hcl, h-val-ome??hcl, acmc-209fna, acmc-209ncl
InChI Key KUGLDBMQKZTXPW-UHFFFAOYSA-N
SMILES Cl.COC(=O)C(N)C(C)C
IUPAC Name methyl 2-amino-3-methylbutanoate;hydrochloride
Molecular Weight (g/mol) 167.63
Quantity 25 g
PubChem CID 12498813
Formula Weight 167.64
Percent Purity 99%
Chemical Name or Material DL-Valine methyl ester hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.