missing translation for 'onlineSavingsMsg'
Learn More

DL-Valine, MP BIOMEDICALS

Product Code. 11457451 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11457451 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11457451 Supplier MP Biomedicals Supplier No. 0210322990
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
  • DL-Valine
  • 99%
  • 500 g

Specifications

CAS 516-06-3
Molecular Weight (g/mol) 117.15
Quantity 500 g
Percent Purity 99%
Chemical Name or Material 2-amino-3-methylbutanoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.