missing translation for 'onlineSavingsMsg'
Learn More

DL-Valine, Thermo Scientific™

Product Code. 15224924 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic Bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15224924 250 g Plastic Bottle 250g
15204924 1 kg Plastic Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15224924 Supplier Thermo Scientific Acros Supplier No. 452832500
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 516-06-3
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00004267
InChI Key KZSNJWFQEVHDMF-UHFFFAOYSA-N
Synonym dl-valine, h-dl-val-oh, valin, valine, dl, dl-2-amino-3-methylbutanoic acid, 2-amino-3-methylbutanoic acid, dl, valina, dl-.alpha.-aminoisovaleric acid, dl-2-amino-3-methylbutyric acid, 2-aminoisovaleric acid
PubChem CID 1182
ChEBI CHEBI:27266
IUPAC Name 2-amino-3-methylbutanoic acid
SMILES CC(C)C(N)C(O)=O

Specifications

Packaging Plastic Bottle
Quantity 250 g
Formula Weight 117.15
Grade Extra Pure
Chemical Name or Material DL-Valine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.