missing translation for 'onlineSavingsMsg'
Learn More

DL-valine, TRC

Product Code. 30454243
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Product Code. Quantity unitSize
30454243 500 mg 500mg
1 options
This item is not returnable. View return policy

Product Code. 30454243

Brand: Toronto Research Chemicals TRCV094195500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DL-Valine
CAS 516-06-3
Molecular Formula C5 H11 N O2
Quantity 500 mg
Synonym Valine, DL-Valine, Valine','DL- (8CI), (RS)-Valine, (±)-2-Amino-3-methylbutyric acid, (±)-Valine, DL-Val, DL-α-Aminoisovaleric acid, NSC 9755
InChI Formula InChI=1S/C5H11NO2/c1-3(2)4(6)5(7)8/h3-4H,6H2,1-2H3,(H,7,8)
SMILES CC(C)C(N)C(=O)O
IUPAC Name 2-amino-3-methylbutanoic acid
Molecular Weight (g/mol) 117.15
Formula Weight 117.079
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.