missing translation for 'onlineSavingsMsg'
Learn More

DMAPT, MedChemExpress

Product Code. 16499537
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16499537 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16499537 Supplier Medchem Express Supplier No. HY16172
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress DMAPT (Dimethylamino Parthenolide), an analogue of Parthenolide (PTL), is an oral active NF-κB inhibitor, with a LD50 of 1.7 μM for cell population in AML cells. Has potential anti-cancer and anti-metastatic effect.

DMAPT (Dimethylamino Parthenolide), an analogue of Parthenolide (PTL), is an oral active NF-κB inhibitor, with a LD50 of 1.7 μM for cell population in AML cells. Has potential anti-cancer and anti-metastatic effect.

Specifications

Chemical Name or Material DMAPT
CAS 870677-05-7
Color White
Purity Grade Notes Research
Molecular Formula C17H27NO3
Quantity 5 mg
Synonym Dimethylamino Parthenolide
Solubility Information DMSO : 125 mg/mL (426.04 mM; Need ultrasonic) ∣H2O : 1.1 mg/mL (3.75 mM; ultrasonic and warming and heat to 40°C)
SMILES C/C1=C/CC[C@]2(C)[C@H](O2)[C@]3([H])[C@](CC1)([H])[C@@H](CN(C)C)C(O3)=O
Molecular Weight (g/mol) 293.4
Formula Weight 293.4
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.