missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DMEM for SILAC

Product Code. 15786803 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 mL
6 x 500 mL
Unit Size:
500mL
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15786803 6 x 500 mL Pack of 6
15645729 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15786803 Supplier Thermo Scientific™ Supplier No. A33822
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Analyze protein expression by mass spectrometry using these media or additives for stable isotope labeling with amino acids in cell culture (SILAC).

Thermo Scientific™ DMEM for SILAC is optimized for use with stable isotope labeling with amino acids in cell culture (SILAC) to analyze protein expression by mass spectrometry (MS).

Features of DMEM for SILAC:

  • Flexible—liquid media deficient in both L-lysine and L-arginine, allowing for more complete proteome coverage through multiple isotopic amino acid labeling
  • High quality—medium is sterile, endotoxin-free, and cell culture-compatible

SILAC is a powerful method to identify and quantify relative differential changes in complex protein samples. This approach entails the in vivo metabolic incorporation of 'heavy' 13C- or 15N-labeled amino acids into proteins followed by mass spectrometry (MS) for the accelerated and comprehensive identification, characterization, and quantitation of proteins.

This SILAC medium is used together with heavy and light amino acids for the differential isotopic labeling of cells for SILAC analysis. SILAC media supplemented with heavy amino acids are capable of nearly 100% label incorporation into proteins of living cells.

Related products

  • NeuCode™ Lysine -080, 50 mg (A33613)
  • Fetal Bovine Serum, dialyzed, US origin (26400036)
  • RPMI 1640 Media for SILAC (88365)
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Mass Spectrometer
Product Type Media
Content And Storage Store at 2°C to 8°C.
Label or Dye Mass Isotope of Amino Acid
Shipping Condition Room Temperature
Starting Material Cell Lysate
Final Product Type Liquid Medium
Quantity 6 x 500 mL
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.