Learn More
Thermo Scientific™ DMF-dG Phosphoramidite, standard grade, serum vial bottle
Shop All Thermo Scientific ProductsDescription
DMF-dG CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxyguanosine (dG) with Dimethylaminoformamidine (DMF) base protection, for use in oligonucleotide synthesis. DNA phosphoramidites are the building blocks for synthesis of DNA or DNA containing oligonucleotides. Each base has unique protecting groups designed specifically to protect the DNA during the chemical synthesis process and produce the highest amount of full length product. Alternative protecting groups on dC (acetyl) and dG (DMF) allow for a faster deprotection of the oligonucleotide. By using Ac-dC, Bz-dA, and DMF-dG monomers, fast cleavage and deprotection can be effected by a 1:1 mixture of aqueous ammonium hydroxide and aqueous methylamine (or AMA) in 10 min. This product is supplied in a 60 mL Amber bottle with a Serum Vial neck style, which is compatible with most ABI Systems|AKTA Systems of oligonucleotide synthesizers.
Features:
- Full chemical name – 5'-O-(4,4'-Dimethoxytrityl)-N2-(dimethylaminoformamidine)-2'-deoxyguanosine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
- Molecular formula – C43H53N8O7P
- Base – dG
- Base Protection – DMF
- Molecular weight – 824.9 g/mol
- CAS number – 330628-04-1
- HPLC Purity – ≥98.0%; M-11 Impurity ≤0.01%
- 31P NMR Purity – ≥98.0%; Sum of non-primary peaks at 140-152 ppm (PIII impurities): ≤1.5%
Specifications
Specifications
| Format | Powder |
| For Use With (Equipment) | AKTA Synthesizers, Applied Biosystems Synthesizers |
| Product Type | DMF-dG CEP |
| Content And Storage | Store in freezer (at or below -20°C). |
| Nucleoside Type | Deoxyguanosine (dG) |
| Reagent Features | Fast deprotection |
| Shipping Condition | Room Temperature |
| Purity | 98% |
| Quantity | 2 g |
| Reagent Type | Phosphoramidite |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.