missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ DMG-PEG 2000

Product Code. 18766165
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18766165 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18766165 Supplier R&D Systems™ Supplier No. 7944/250
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. PEGylated myristoyl diglyceride; commonly used in the formation of lipid nanoparticles (LNPs)

DMG-PEG 2000 is a PEGylated myristoyl diglyceride consisting of one unbranched PEG 2000 chain and two saturated fatty acid chains (C14:0) covalently attached to a glycerol. DMG-PEG 2000 is commonly used in the formation of lipid nanoparticles (LNPs) and liposomes to prevent serum protein adsorption, nanoparticle aggregation and increase in vivo circulation time. DMG-PEG 2000 is a component of LNPs which can be used for delivery of RNA-based vaccines or mRNA therapeutics.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2-Dimyristoyl-rac-glycero-3-methoxypolyethylene glycol-2000
CAS 160743-62-4
Quantity 250 mg
Product Line Tocris
Target Lipid Nanoparticles
Molecular Formula (C2H4O)nC32H62O5
Purity 0.9
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.