missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ DMH-1

Product Code. 18005362
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18005362 10 mg 10mg
18065942 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18005362 Supplier R&D Systems™ Supplier No. 4126/10
  • ONLINE EXCLUSIVE PRICE
    338.00 €
    319.20 € / 10mg
    Save 18.80 €
    5% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective ALK2 inhibitor

DMH-1 is a selective inhibitor of bone morphogenic protein (BMP) type-I receptor activin receptor-like kinase 2 (ALK2) receptor (IC50 = 108 nM or 12.6 nM in in vitro kinase assays). DMH-1 exhibits 6- and 19-fold selectivity for ALK-2 over ALK-1 and ALK-3, respectively, and no significant inhibition of AMPK, ALK5, KDR (VEGFR-2) or PDGFRβ receptors. DMH-1 blocks BMP4-induced phosphorylation of Smads 1, 5 and 8 in HEK293 cells. Promotes neurogenesis in human induced pluripotent stem cells (iPSCs) when used in combination with SB 431542 (Cat. No. 1614). DMH-1 suppresses lung cancer cell proliferation, migration, invasion in vitro and reduces tumor growth in a mouse lung cancer xenograft model. DMH-1 inhibits cellular autophagy responses. DMH-1 induces intestinal differentiation in human intestinal organoids (hIOs) derived from human pluripotent stems cells (hPSCs).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1206711-16-1
Molecular Formula C24H20N4O

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 380.44
Purity >98%
Molecular Weight (g/mol) 380.45
Chemical Name or Material DMH-1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.