missing translation for 'onlineSavingsMsg'
Learn More

DMH-1, MedChemExpress

Product Code. 16475048
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16475048 10 mg 10mg
16451287 50 mg 50mg
16415796 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16475048 Supplier Medchem Express Supplier No. HY12273
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress DMH-1 is a potent and selective BMP inhibitor with IC50s of 27/107.9/<5/47.6 nM for ALK1/ALK2/ALK3/ALK6, respectively.

DMH-1 is a potent and selective BMP inhibitor with IC50s of 27/107.9/<5/47.6 nM for ALK1/ALK2/ALK3/ALK6, respectively.

Specifications

Chemical Name or Material DMH-1
CAS 1206711-16-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C24H20N4O
Quantity 10 mg
Solubility Information DMSO : 11.5 mg/mL (30.23 mM; Need ultrasonic and warming)
SMILES CC(C)OC1=CC=C(C2=CN3C(N=C2)=C(C4=CC=NC5=CC=CC=C45)C=N3)C=C1
Molecular Weight (g/mol) 380.44
Formula Weight 380.44
Percent Purity 99.58%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.