missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies DMH1
Description
- DMH1 (dorsomorphin homolog 1) is a selective inhibitor of activin receptor-like kinase 2 (ALK2; IC50 = 13 - 108 nM), a type I bone morphogenetic protein (BMP) receptor (Hao et al.; Mohedas et al.) DMH1 exhibits no detectable inhibition of ALK4, ALK5, AMPK, KDR (VEGFR2), or PDGFRβ, although it inhibits ALK1 and ALK3 at nanomolar concentrations (Hao et al.; Mohedas et al.)
Specifications
Specifications
| CAS | 1206711-16-1 |
| Quantity | 10 mg |
| Molecular Formula | C24H20N4O |
| Purity | ≥98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?