Learn More
Tocris Bioscience™ DMXAA
mSTING agonist; induces antitumor immunological responses
Brand:  Tocris Bioscience™ 5601/50
Encompass
Product Code. 15726966
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
DMXAA is a mSTING agonist; selective for mouse STING over human STING. Induces IFN-β and cytokine production from bone marrow-derived dendritic cells. Causes tumor regression following intratumoral administration in mouse tumor models. Induces an adaptive immune response to reduce systemic tumor growth and provides immunological memory against autologous tumor re-challenge. Also antiviral.
Specifications
Specifications
5,6-Dimethyl-9-oxo-9 H-xanthene-4-acetic acid
117570-53-3