missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ DMXAA
GREENER_CHOICE

Product Code. 18702138
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18702138 10 mg 10mg
18718688 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18702138 Supplier Tocris Bioscience™ Supplier No. 5601/10
  • ONLINE EXCLUSIVE PRICE
    221.00 €
    208.95 € / 10mg
    Save 12.05 €
    5% Off
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

mSTING agonist; induces antitumor immunological responses

DMXAA is a mSTING agonist; selective for mouse STING over human STING. Induces IFN-β and cytokine production from bone marrow-derived dendritic cells. Causes tumor regression following intratumoral administration in mouse tumor models. Induces an adaptive immune response to reduce systemic tumor growth and provides immunological memory against autologous tumor re-challenge. Also antiviral.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5,6-Dimethyl-9-oxo-9 H-xanthene-4-acetic acid
CAS 117570-53-3
Quantity 10 mg
Target STING Agonists
Molecular Formula C17H14O4
Formula Weight 282.29
Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.