missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ DMXAA
GREENER_CHOICE

Product Code. p-7110965
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18702138

Brand: Tocris Bioscience™ 5601/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

mSTING agonist; induces antitumor immunological responses

DMXAA is a mSTING agonist; selective for mouse STING over human STING. Induces IFN-β and cytokine production from bone marrow-derived dendritic cells. Causes tumor regression following intratumoral administration in mouse tumor models. Induces an adaptive immune response to reduce systemic tumor growth and provides immunological memory against autologous tumor re-challenge. Also antiviral.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5,6-Dimethyl-9-oxo-9 H-xanthene-4-acetic acid
CAS 117570-53-3
Quantity 10 mg
Target STING Agonists
Molecular Formula C17H14O4
Formula Weight 282.29
Purity 99%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.