missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ DMXAA
Description
DMXAA is a mSTING agonist; selective for mouse STING over human STING. Induces IFN-β and cytokine production from bone marrow-derived dendritic cells. Causes tumor regression following intratumoral administration in mouse tumor models. Induces an adaptive immune response to reduce systemic tumor growth and provides immunological memory against autologous tumor re-challenge. Also antiviral.
Specifications
Specifications
| Chemical Name or Material | 5,6-Dimethyl-9-oxo-9 H-xanthene-4-acetic acid |
| CAS | 117570-53-3 |
| Quantity | 50 mg |
| Product Line | Tocris |
| Target | STING Agonists |
| Molecular Formula | C17H14O4 |
| Formula Weight | 282.29 |
| Purity | 99% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?