missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human DNA-PK Native Protein

Product Code. 15378517
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
Unit Size:
100µg
10µg
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15378517 1 mg Each
15117975 100 μg 100µg
15428803 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15378517 Supplier Invitrogen™ Supplier No. PV5865

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Native Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Native human protein purified from MOLT-4 cells. No special measures were taken to activate this kinase. Protein Form: Full Length.

DNA-PK is a heterotrimeric, DNA-dependent protein kinase that initiates signaling cascades in response to DNA damage. DNA-PK is an atypical kinase, closely related to the PI3 kinases, and is often potently inhibited by small molecules designed to target PI3 kinases. DNA-PK encodes a component of the mediator complex, a transcriptional coactivator complex thought to be required for the expression of almost all genes. The mediator complex is recruited by transcriptional activators or nuclear receptors to induce gene expression, possibly by interacting with RNA polymerase II and promoting the formation of a transcriptional pre-initiation complex. Multiple transcript variants encoding different isoforms have been found for DNA-PK.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P78527
Concentration Conc. Not Determined
For Use With (Application) Kinase Assay
Formulation 25mM HEPES with 10mM MgCl2, 50mM KCl, 1mM DTT, 0.2mM EDTA, 10% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 5591
Molecular Weight (g/mol) 600 kDa
Name Human DNA-PK
pH Range 7.5
Quantity 1 mg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias AI326420; AU019811; DNA-dependent protein kinase catalytic subunit; DNAPDcs; DNAPK; DNA-PK catalytic subunit; DNA-PKcs; DNPK1; DOXNPH; doxorubicin nephropathy; dxnph; hyper-radiosensitivity of murine scid mutation, complementing 1; HYRC; HYRC1; I79_017613; IMD26; p350; p460; Prkdc; protein kinase, DNA activated, catalytic polypeptide; protein kinase, DNA-activated, catalytic polypeptide; scid; severe combined immunodeficiency; slip; Xrcc7
Common Name DNA-PK
Gene Symbol PRKDC
Kinase Family PIKK Kinase Family
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Native
Protein Tag Untagged
Sequence Full length
Content And Storage Kinase proteins are shipped on dry ice and should be stored at -80°C.
Kinase Group Ser⁄Thr Kinases: Atypical
Shipping Condition Dry Ice
Expression System Baculovirus
Protein Family Kinases & Inhibitors
Protein Form Full Length
Form Liquid
Protein Subtype Serine/Threonine Kinases, Atypical
Research Category Drug Discovery: Signal Transduction
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.