missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ DNA Set for NucleoBond™ RNA Soil Mini

Product Code. 16331606
Change view
Click to view available options
No. of Reactions:
10
50
Unit Size:
10 reactions
50 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
16331606 50 50 reactions
16321606 10 10 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16331606 Supplier Macherey-Nagel™ Supplier No. 740143.50
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Set for parallel DNA isolation from soil samples with NucleoBond RNA Soil Mini, NucleoSpin DNA Columns included.

  • DNA Elution Set (NucleoBond™ RNA Soil Mini kit required)
  • Parallel preparation of RNA and DNA from soil samples

Specifications

Format Kit
Isolation Technology Silica Membrane
Sample Type Soil
For Use With (Equipment) NucleoBond™ RNA Soil Kit
For Use With (Application) RNA Purification
No. of Reactions 50
Sample Size 1-2 g
Test Time 15 min / 12 preps
Yield 5-50 μg

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.