Learn More
STEMCELL Technologies DNase I
Product Code. p-7217577
Encompass_Preferred
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Product Code. 15964601
Brand: STEMCELL Technologies 07469
For digestion of DNA
  • Deoxyribonuclease I (DNase I) is an endonuclease consisting of a single glycosylated polypeptide chain with two disulfide bonds
  • DNase is often included in tissue dissociation protocols to digest DNA that has leaked into the dissociation medium as a result of cell damage
  • DNase I preferentially cleaves phosphodiester linkages adjacent to pyrimidine nucleotides in both single- and double-stranded DNA, yielding polynucleotides with 5'-phosphate and 3'-hydroxyl groups (Bernardi et al.) DNase I has been used for the digestion of human tissues such as microglia (Klegeris & McGeer), cartilage (Dunham & Koch), colon (Fukushima & Fiocchi), epithelium (Fukushima & Fiocchi), liver (Vatakis et al.), lung (Fujino et al.), neural (Fuja et al.), and stem cells (Kusuma et al.)
Specifications
For Use With (Application) Digestion of DNA
Quantity 25 mg