missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DNase/RNase-Free Deionized Water

Product Code. 30826300 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 L
2 L
Unit Size:
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30826300 2 L -
17856196 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30826300 Supplier Thermo Scientific™ Supplier No. A57774
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

DNase/RNase-Free Deionized Water for use in all molecular biology applications.

DNase/RNase-Free Deionized Water is designed for use in all molecular biology applications and provided in a convenient 1-liter and 2-liter bottles.

Confidence in your applications

The water is 0.22-micron-filtered to remove bacteria, most viruses, and other particulates, making it suitable for preparation of growth media. Water quality is monitored with extensive testing for the presence of DNA/RNA nucleases, protease activity, and human and bacterial DNA.

Applications

DNase/RNase-Free Deionized Water is suitable for various molecular biology applications, including PCR, qPCR, cDNA synthesis, cloning, next-generation sequencing, nucleic acid extraction or storage, and research applications involving low amounts of nucleic acids. Additionally, it can be used for applications that are sensitive to the presence of ions, like solution or buffer preparation, mass spectrometry, and spectrophotometry.

Manufacturing excellence

Manufactured by Thermo Fisher Scientific Baltics UAB in compliance with ISO 9001 and ISO 13485 certified quality management system.

TRUSTED_SUSTAINABILITY

Specifications

Synonym Nuclease-free deionized water
Grade Molecular Biology
Treatment(s) Not DEPC-Treated
Quantity 2 L

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.