missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ DNQX disodium salt
GREENER_CHOICE

Product Code. p-4783917
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001300

Brand: Tocris Bioscience™ 2312/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective non-NMDA iGluR antagonist; water-soluble salt of DNQX (Cat. No. 0189)

DNQX disodium salt is a more water-soluble form of DNQX (Cat. No. 0189). DNQX is a selective non-NMDA glutamate receptor antagonist. (IC50 = 0.1 μM and 0.5 μM for kainate and AMPA receptors, respectively). DNQX is a neuroleptic agent that shows pro-oxidative properties. DNQX selectively depolarizes rat thalamic reticular nucleus neurons.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1312992-24-7
Molecular Formula C8H2N4Na2O6

Specifications

Quantity 10 mg
Formula Weight 296.1
Purity >98%
Molecular Weight (g/mol) 296.106
Chemical Name or Material DNQX disodium salt
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.