missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Dobutamine hydrochloride

Product Code. 30001096
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001096 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001096 Supplier R&D Systems™ Supplier No. 0515/50
  • ONLINE EXCLUSIVE PRICE
    325.35 €
    292.95 € / 50mg
    Save 32.40 €
    9% Off
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. α1, β1 and β2 agonist

β1- and β2-adrenoceptor agonist with some action at the α1-adrenoceptor.
TRUSTED_SUSTAINABILITY

Specifications

CAS 49745-95-1
Quantity 50 mg
Product Line Tocris
Molecular Formula C18H24ClNO3
Formula Weight 337.85
Purity >99%
Molecular Weight (g/mol) 337.84
Chemical Name or Material Dobutamine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.