missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Docetaxel

Product Code. 18782674
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18782674 10 mg 10mg
18732704 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18782674 Supplier R&D Systems™ Supplier No. 4056/10
  • ONLINE EXCLUSIVE PRICE
    225.08 €
    202.65 € / 10mg
    Save 22.43 €
    9% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Microtubule stabilizer

Binds and stabilizes microtubules; leads to cell cycle arrest and apoptosis. Displays significant antitumor activity in ovarian and gastric tumors. Prevents microtubule depolymerization and disassembly in the absence of GTP. Exhibits cytotoxic activity.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114977-28-5
Molecular Formula C43H53NO14

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 807.88
Purity >99%
Molecular Weight (g/mol) 807.89
Chemical Name or Material Docetaxel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.