missing translation for 'onlineSavingsMsg'
Learn More

Docosahexaenoic Acid, TRC

Product Code. 30008563
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008563 250 mg 250mg
30006410 50 mg 50mg
17910105 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008563 Supplier Toronto Research Chemicals Supplier No. TRCD494500250MG
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6217-54-5
Molecular Formula C22 H32 O2
Molecular Weight (g/mol) 328.49
Synonym 4,7,10,13,16,19-Docosahexaenoic acid, (4Z,7Z,10Z,13Z,16Z,19Z)-, 4,7,10,13,16,19-Docosahexaenoic acid, (all-Z)- (8CI), Docosahexaenoic acid (6CI), (4Z,7Z,10Z,13Z,16Z,19Z)-4,7,10,13,16,19-Docosahexaenoic acid, (4Z,7Z,10Z,13Z,16Z,19Z)-Docosahexaenoic acid, (4Z,7Z,10Z,13Z,16Z,19Z)-Docosahexenoic acid, (all-Z)-4,7,10,13,16,19-Docosahexaenoic acid, 4-cis,7-cis,10-cis,13-cis,16-cis,19-cis-Docosahexaenoic acid, AlgaMac 3050, Algatrium, AquaGrow Advantage, C22:6, Cervonic acid, DHA, DHA 22, DHA 22HG, DHA 46, DHA 46A, DHA S Rosemary Sun, DHASCO, DHASCO-HM, DHASCO-S, DHASCO-T, Doconexent, Efalex, Marinol D 50TG, Martek DHA HM, Martek DHA Powder KS 35, Martek DHA Powder KSF 58, Martek DHA-S, Maxigard, Monolife 50, Monolife 90, Ropufa 60, S.Presso, all-Z-Docosahexaenoic acid, all-cis-4,7,10,13,16,19-Docosahexaenoic acid, δ4,7,10,13,16,19-Docosahexaenoic acid
IUPAC Name (4Z,7Z,10Z,13Z,16Z,19Z)-docosa-4,7,10,13,16,19-hexaenoic acid
SMILES CC\C=C/C\C=C/C\C=C/C\C=C/C\C=C/C\C=C/CCC(=O)O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C22H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22(23)24/h3-4,6-7,9-10,12-13,15-16,18-19H,2,5,8,11,14,17,20-21H2,1H3,(H,23,24)/b4-3-,7-6-,10-9-,13-12-,16-15-,19-18-
Formula Weight 328.2402
Chemical Name or Material cis-Docosahexaenoic Acid (DHA)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.