missing translation for 'onlineSavingsMsg'
Learn More

Docosanoic acid, MedChemExpress

Product Code. 16479366
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16479366 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16479366 Supplier Medchem Express Supplier No. HYW013049
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Docosanoic acid is poorly absorbed, and a cholesterol-raising saturated fatty acid in humans.

Docosanoic acid is poorly absorbed, and a cholesterol-raising saturated fatty acid in humans.

Specifications

Chemical Name or Material Docosanoic acid
CAS 112-85-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H44O2
Quantity 100 mg
Solubility Information DMSO : 1.43 mg/mL (4.20 mM; ultrasonic and warming and heat to 80°C) ∣H2O : 1 mg/mL (2.94 mM; ultrasonic and warming and heat to 80°C)
SMILES CCCCCCCCCCCCCCCCCCCCCC(O)=O
Molecular Weight (g/mol) 340.59
Formula Weight 340.59
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.