Learn More
Vilbert Lourmat™ Docprint CX5 Gel Documentation System
Product Code. 11766568
Change view
Click to view available options
Product Type: Gel Documentation System
Product Code.
11766568 Product Type: Gel Documentation System
1 options
Product Code. 11766568 Supplier Vilbert Lourmat™ Supplier No. 111101121
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Complete solution without a PC, this robust Doc-Print VX2 Standalone System is ideal for the documentation of electrophoresis gels, and membranes.
  • For electrophoresis gels, autoradiographs, membranes
  • Complete solution without PC
  • Image transfer via USB key
  • Self-contained system for simple documentation of gels
  • Control unit with 8 in. screen for viewing images and thermoformed ABS camera support cone
  • 12-bit scientific CCD camera (4096 levels of gray)
  • Modulating exposure time, indication of saturation of the spots on the display
  • Large 8 TFT LCD screen
  • Save images to USB flash drive
  • Possibility to connect directly a thermal printer or USB inkjet
  • Software provided for editing, publication analysis: molecular weight calculation and optical density, inlay of image comments
Specifications
Model Docprint CX5 Gel
Packaging Quantity 1Each
Product Type Gel Documentation System