missing translation for 'onlineSavingsMsg'
Learn More

Docusate Sodium, MedChemExpress

Product Code. 16408716
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16408716 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16408716 Supplier Medchem Express Supplier No. HYB1268
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Docusate Sodium (Dioctyl sulfosuccinate sodium salt) is a laxative used to for the research of constipation, for constipation due to the use of opiates it maybe used with a stimulant laxative, can be taken by mouth or rectally.

Docusate Sodium (Dioctyl sulfosuccinate sodium salt) is a laxative used to for the research of constipation, for constipation due to the use of opiates it maybe used with a stimulant laxative, can be taken by mouth or rectally.

Specifications

Chemical Name or Material Docusate Sodium
CAS 577-11-7
Color White
Purity Grade Notes Research
Molecular Formula C20H37NaO7S
Quantity 100 mg
Synonym Dioctyl sulfosuccinate sodium salt
Solubility Information H2O : ≥ 20 mg/mL (44.99 mM)
SMILES O=C(OCC(CC)CCCC)C(S(=O)(O[Na])=O)CC(OCC(CC)CCCC)=O
Molecular Weight (g/mol) 444.56
Formula Weight 444.56
Percent Purity 98.0%
Grade Research
Physical Form Oil
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.