missing translation for 'onlineSavingsMsg'
Learn More

Dodecacarbonyltriosmium, 99%

Product Code. 11392118
Change view
Click to view available options
Quantity:
0.5 g
2 g
Unit Size:
0.5g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11392118 0.5 g 0.5g
11302128 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392118 Supplier Thermo Scientific Alfa Aesar Supplier No. 013038.02
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 15696-40-9 | C12O12Os3 | 906.81 g/mol

Dodecacarbonyltriosmium is an important precursor to organo-osmium compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15696-40-9
Molecular Formula C12O12Os3
Molecular Weight (g/mol) 906.81
MDL Number MFCD00011149
InChI Key VUBLMKVEIPBYME-UHFFFAOYSA-N
Synonym osmium carbonyl, triangulo-dodecacarbonyltriosmium, triosmium dodecacarbonyl, tri-osmium dodecacarbonyl
PubChem CID 6096995
IUPAC Name carbon monoxide;osmium
SMILES [C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Os].[Os].[Os]

Specifications

Melting Point 224°C
Quantity 0.5 g
UN Number UN3466
Solubility Information Insoluble in water. Slightly soluble in organic solvents.
Formula Weight 906.73
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material Dodecacarbonyltriosmium

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.