missing translation for 'onlineSavingsMsg'
Learn More

Dodecane, ≥99%, Reagent Grade, Honeywell™

Product Code. p-7120396 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
100mL
2.5 L
500mL
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 15607810

Brand: Honeywell Research Chemicals D221104100ML

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-40-3
Molecular Formula C12H26
Molecular Weight (g/mol) 170.34
MDL Number MFCD00008969
InChI Key SNRUBQQJIBEYMU-UHFFFAOYSA-N
Synonym n-dodecane, dihexyl, bihexyl, adakane 12, n-dodecan, n-dodecan german, duodecane, alkanes, c10-14, undecane, methyl, ccris 661
PubChem CID 8182
ChEBI CHEBI:28817
IUPAC Name dodecane
SMILES CCCCCCCCCCCC

Specifications

Melting Point -9.6°C
Color Colorless
Density 0.75g/mL at 25°C
Boiling Point 215°C to 217°C
Odor Characteristic
Packaging Glass bottle
Linear Formula CH3(CH2)10CH3
Quantity 100 mL
Beilstein 1697175
Vapor Density 5.96 (vs air)
Formula Weight 170.33g/mol
Percent Purity ≥99%
Chemical Name or Material Dodecane
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.