missing translation for 'onlineSavingsMsg'
Learn More

Dodecane, TRC

Product Code. 30136600
Change view
Click to view available options
Quantity:
50 mL
100 mL
150 mL
Unit Size:
100mL
150mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136600 150 mL 150mL
30136776 50 mL 50mL
17913455 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 30136600 Supplier Toronto Research Chemicals Supplier No. TRCD494625150ML
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-40-3
Molecular Formula C12 H26
Molecular Weight (g/mol) 170.33
Synonym Dodecane, Adakane 12, Ba 51-090453, C12-n-Alkane, Cactus Normal Paraffin N 12D, NSC 8714, Parafol 12-97, Parafol C12-97, n-Dodecane
IUPAC Name dodecane
SMILES CCCCCCCCCCCC

Specifications

Quantity 150 mL
InChI Formula InChI=1S/C12H26/c1-3-5-7-9-11-12-10-8-6-4-2/h3-12H2,1-2H3
Formula Weight 170.2035
Chemical Name or Material n-Dodecane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.