missing translation for 'onlineSavingsMsg'
Learn More

Dodecane, TRC

Product Code. 30136776
Change view
Click to view available options
Quantity:
50 mL
100 mL
150 mL
Unit Size:
100mL
150mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136776 50 mL 50mL
30136600 150 mL 150mL
17913455 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 30136776 Supplier Toronto Research Chemicals Supplier No. TRCD49462550ML
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-40-3
Molecular Formula C12 H26
Molecular Weight (g/mol) 170.33
Synonym Dodecane, Adakane 12, Ba 51-090453, C12-n-Alkane, Cactus Normal Paraffin N 12D, NSC 8714, Parafol 12-97, Parafol C12-97, n-Dodecane
IUPAC Name dodecane
SMILES CCCCCCCCCCCC

Specifications

Quantity 50 mL
InChI Formula InChI=1S/C12H26/c1-3-5-7-9-11-12-10-8-6-4-2/h3-12H2,1-2H3
Formula Weight 170.2035
Chemical Name or Material n-Dodecane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.