missing translation for 'onlineSavingsMsg'
Learn More

Dodecanoic acid, typically 99.5%

Product Code. 11379697
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11379697 250 g 250g
11359697 50 g 50g
11369697 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11379697 Supplier Thermo Scientific Alfa Aesar Supplier No. 042038.30
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 143-07-7 | C12H24O2 | 200.322 g/mol

Dodecanoic acid is used in the preparation of cosmetics, soaps, alkyd resins and wetting agents. It is also used to measure the molar mass of an unknown substance through freezing point depression. It is also used as a food additive and an active component in a treatment for acne. In addition to this, it is a substrate for acylation of certain proteins based on the murine studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-07-7
Molecular Formula C12H24O2
Molecular Weight (g/mol) 200.322
MDL Number MFCD00002736
InChI Key POULHZVOKOAJMA-UHFFFAOYSA-N
Synonym lauric acid, n-dodecanoic acid, dodecylic acid, vulvic acid, laurostearic acid, dodecoic acid, duodecylic acid, 1-undecanecarboxylic acid, aliphat no. 4, neo-fat 12
PubChem CID 3893
ChEBI CHEBI:30805
IUPAC Name dodecanoic acid
SMILES CCCCCCCCCCCC(=O)O

Specifications

Melting Point 43°C to 45°C
Density 0.883 g/mL
Boiling Point 180°C (21 mmHg)
Flash Point 165°C (329°F)
Odor Odorless
Quantity 250 g
Beilstein 1099477
Merck Index 14,5384
Solubility Information Soluble in water,benzene,acetone,alcohol,petroleum ether,dimethyl sulfoxide and dimethyl formamide. Slightly soluble in chloroform.
Formula Weight 200.32
Percent Purity 99.5% (typically)
Physical Form Crystalline
Chemical Name or Material Dodecanoic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.