missing translation for 'onlineSavingsMsg'
Learn More

Dodecanoyl chloride, 98%

Product Code. 11440873
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11440873 500 g 500g
11430873 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11440873 Supplier Thermo Scientific Alfa Aesar Supplier No. A13294.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 112-16-3 | C12H23ClO | 218.77 g/mol

Lauroyl chloride is used as tailoring agent for chemical modification of nanocelluloses of different length, nanofibrillated cellulose and cellulose nanocrystals and in the preparation of acylated collagen with water solubility and better surface activity. It is also employed as organic low-friction boundary lubricant in the preparation of novel polyvinyl alcohol hydrogel.It is used in the synthesis of hemicellulose-based hydrophobic biomaterials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lauroyl chloride is used as tailoring agent for chemical modification of nanocelluloses of different length, nanofibrillated cellulose and cellulose nanocrystals and in the preparation of acylated collagen with water solubility and better surface activity. It is also employed as organic low-friction boundary lubricant in the preparation of novel polyvinyl alcohol hydrogel.It is used in the synthesis of hemicellulose-based hydrophobic biomaterials.

Solubility
Soluble in ethanol and methanol.

Notes
Moisture sensitive. Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-16-3
Molecular Formula C12H23ClO
Molecular Weight (g/mol) 218.77
MDL Number MFCD00000740
InChI Key NQGIJDNPUZEBRU-UHFFFAOYSA-N
Synonym lauroyl chloride, lauroylchloride, n-dodecanoyl chloride, lauric acid chloride, dodecanoic acid, chloride, unii-9lhl10777i, laurel acetyl chlorine, dodecanoylchloride, lauric acid, chloride, pubchem21365
PubChem CID 8166
IUPAC Name dodecanoyl chloride
SMILES CCCCCCCCCCCC(Cl)=O

Specifications

Melting Point -17°C
Density 0.922
Boiling Point 130°C to 132°C (7 mmHg)
Flash Point 140°C (284°F)
Linear Formula CH3(CH2)10COCl
Refractive Index 1.446
Quantity 500 g
UN Number UN3265
Beilstein 1281201
Sensitivity Moisture sensitive
Solubility Information Soluble in ethanol and methanol.
Formula Weight 218.77
Percent Purity 98%
Chemical Name or Material Dodecanoyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.