Learn More
Thermo Scientific Chemicals Dodecyl sulfate, sodium salt, 98%, for biochemistry, suitable for electrophoresis
Brand: Thermo Scientific Chemicals 419530010
Code : 29
Additional Details : CAS Number : 151-21-3 Weight : 1.00000kg Transport : UN number : 1325 Chem class : 4.1 Pack group : III
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For biochemistrySpecifications
Dodecyl sulfate, sodium salt | |
151-21-3,7757-82-6,7647-14-5 | |
Fine Crystalline, Flaky or Granular Powder | |
98% | |
CH3(CH2)11OSO3Na | |
01, III, 1786 | |
>150°C | |
sodium dodecyl sulfate, sodium lauryl sulfate, sodium dodecylsulfate, sodium lauryl sulphate, sodium dodecyl sulphate, dodecyl sulfate, sodium salt, anticerumen, gardinol, neutrazyme, duponal | |
DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
sodium dodecyl sulfate | |
3423265 | |
288.38 | |
Biochemical | |
1% max. | |
3% aq. soln. clear, colorless. 1% soln. in EtOH clear, colorless |
206.0°C | |
White | |
1 kg | |
C12H25NaO4S | |
MFCD00036175 | |
13,281 | |
15, 8768 | |
Solubility in water: 130g/L (20°C). Other solubilities: partly soluble in alcohol, soluble 75g/L in ethanol (20°C) | |
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O | |
288.38 | |
CHEBI:8984 | |
98% | |
Authentic | |
Plastic bottle |
Safety and Handling
- Dodecyl sulfate sodium salt
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Flammable solid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H228-Flammable solid.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only