missing translation for 'onlineSavingsMsg'
Learn More

Dodecyl Trimethyl Ammonium Bromide, Ion Pair Chromatography, Fisher Chemical

Product Code. 10696252 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Plastic powder jar
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10696252 25 g Plastic powder jar 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10696252 Supplier Fisher Chemical Supplier No. D/5311/46
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 246°C
CAS 1119-94-4
Color White
Physical Form Powder/Solid
Quantity 25 g
Packaging Plastic powder jar
Molecular Formula C15H34BrN
MDL Number 11767
Synonym dodecyltrimethylammonium bromide, dtab, dctab, ltab, lauryltrimethylammonium bromide, n,n,n-trimethyldodecan-1-aminium bromide, laurtrimonium bromide, dodecyl trimethyl ammonium bromide, morpan d, 1-dodecanaminium, n,n,n-trimethyl-, bromide
InChI Key XJWSAJYUBXQQDR-UHFFFAOYSA-M
SMILES CCCCCCCCCCCC[N+](C)(C)C.[Br-]
IUPAC Name dodecyl(trimethyl)azanium;bromide
Molecular Weight (g/mol) 308.348
PubChem CID 14249
ChEBI CHEBI:282662
Formula Weight 308.34g/mol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dodecyltrimethylammonium bromide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.