missing translation for 'onlineSavingsMsg'
Learn More

Dodecylbenzene, TRC

Product Code. 30361098
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30361098 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30361098 Supplier Toronto Research Chemicals Supplier No. TRCD12331025G
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dodecylbenzene
CAS 123-01-3
Molecular Formula C18 H30
Quantity 25 g
Synonym Dodecane, 1-phenyl- (8CI), Dodecylbenzene, 1-Dodecylbenzene, 1-Phenyl-n-dodecane, 1-Phenyldodecane, Alkylate P 1, Detergent Alkylate No. 2, Laurylbenzene, NSC 102805, NSC 4584, Nalkylene 500, n-Dodecylbenzene
InChI Formula InChI=1S/C18H30/c1-2-3-4-5-6-7-8-9-10-12-15-18-16-13-11-14-17-18/h11,13-14,16-17H,2-10,12,15H2,1H3
SMILES CCCCCCCCCCCCc1ccccc1
IUPAC Name dodecylbenzene
Molecular Weight (g/mol) 246.43
Formula Weight 246.2348
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.