missing translation for 'onlineSavingsMsg'
Learn More

Dodecylbenzenesulfonic acid sodium salt, ≥80%, MP Biomedicals™

Product Code. 11471261 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11471261 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11471261 Supplier MP Biomedicals Supplier No. 0215788991
 Restricted Item
Estimated Shipment: 29-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A member of the linear alkylbenzenesulfonates

Specifications

CAS 25155-30-0
Color White
Assay Percent Range ≥80%
Molecular Formula C18H29NaO3S
MDL Number MFCD00011508
Quantity 1 kg
Synonym Sodium Dodecylbenzenesulfonate, Abeson Nam
InChI Key HFQQZARZPUDIFP-UHFFFAOYSA-M
Molecular Weight (g/mol) 348.48
Formula Weight 348.5
Physical Form Flakes
Chemical Name or Material Dodecylbenzenesulfonic acid sodium salt
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dodecylbenzene Sulfonic Acid Sodium Salt
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P330-Rinse mouth.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: dodecylbenzene sulphonic acid sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.